Rapamycin ≥98%

Supplier: Ambeed
Warning

Synonyms: (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone

A656002-1G A656002-25MG A656002-100MG A656002-250MG A656002-5G
103799-710EA 15 USD
103799-710 103840-416 77573-106 77573-108 77594-996 77614-024 77614-026
Rapamycin ≥98%
Rapamycin

Caution: Sealed in dry, store in freezer, under –20 °C.

Formula: C₅₁H₇₉NO₁₃
Boiling Pt: 973 °C (760 mm Hg)
Flash Pt: 542.3 °C
Storage Temperature: Freezer
MDL Number: MFCD00867594
CAS Number: 53123-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR