Trypan blue 70% (dye content)

Supplier: Ambeed
Danger

Synonyms: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulfonic acid tetrasodium salt, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulfonate), AzidinBlue, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulfonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)

A621935-5G
77613-932EA 15.45 USD
77613-932
Trypan blue 70% (dye content)
Trypan Blue

Caution: for Research Use Only

Formula: C₃₄H₂₄N₆Na₄O₁₄S₄
MW: 960.82 g/mol
MDL Number: MFCD00003969
CAS Number: 72-57-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR