2,7-Diamino-9,9-di-n-octylfluorene 98%
Supplier: Ambeed
|
Warning
|
A391620-250MG
A391620-1G
A391620-100MG
77016-194EA
23
USD
77016-194
77613-182
77890-806
2,7-Diamino-9,9-di-n-octylfluorene 98%
2,7-Diamino-9,9-di-n-octylfluorene
Caution: for Research Use Only
Formula:
C₂₉H₄₄N₂ |
MDL Number:
MFCD27923070 CAS Number: 851042-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...