Fmoc-Dap(Alloc)-OH 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Dap(Aloc)-OH, (S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid
A210001-1G
A210001-250MG
A210001-5G
A210001-10G
A210001-25G
A210001-100G
77097-638EA
23
USD
77097-638
77097-640
77097-642
77568-024
77612-060
77685-545
Fmoc-Dap(Alloc)-OH 95%
Fmoc-Dap(Alloc)-OH
Caution: For research use only.
Formula:
C₂₂H₂₂N₂O₆ |
MDL Number:
MFCD00273468 CAS Number: 188970-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...