5,15-Diphenylporphyrin ≥90%

Supplier: Ambeed
Warning

Synonyms: 5,15-Diphenyl-21H,23H-porphine

A180110-250MG
77611-746EA 225.57 USD
77611-746
5,15-Diphenylporphyrin ≥90%
5,15-Diphenylporphyrin

Caution: for Research Use Only

Formula: C₃₂H₂₂N₄
MW: 462.56 g/mol
MDL Number: MFCD08276335
CAS Number: 22112-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR