L(-)-Carnitine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Levocarnitine hydrochloride, L-Carnitine hydrochloride, (R)-3-Carboxy-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride, LC-80, (R)-3-Hydroxy-4-(trimethylammonio)butyrate hydrochloride, L-Carnitine chloride, (R)-Carnitine hydrochloride, Levocarnitine chloride

A142902-100G
103808-634EA 171 USD
103808-634 103834-256 77611-312
L(-)-Carnitine hydrochloride 97%
L(-)-Carnitine hydrochloride

Caution: Keep in a dark place, sealed in dry, room temperature

Formula: C₇H₁₆ClNO₃
Melting Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD00066100
CAS Number: 6645-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR