Imperatorin 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-((3-Methylbut-2-en-1-yl)oxy)-7H-furo[3,2-g]chromen-7-one
A118402-10MG
A118402-1MG
A118402-25MG
A118402-5MG
A118402-1G
A118402-250MG
77094-342EA
21
USD
77094-342
77094-344
77094-346
77094-348
77610-970
77685-045
Imperatorin 97%
Imperatorin
Caution: for Research Use Only
Formula:
C₁₆H₁₄O₄ |
MDL Number:
MFCD00016881 CAS Number: 482-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...