SafeMask® Premier Elite Proshield™ Procedure Earloop Face Masks with Visor
Supplier: Medicom
2025
77610-650BX 23.85 USD
77610-650
SafeMask® Premier Elite Proshield™ Procedure Earloop Face Masks with Visor
Masks Surgical Masks
Procedure masks are intended to be worn by personnel during surgical or other medical procedures to protect both the patient and the operating personnel from the transfer of microorganisms, body fluids, and particulate material.
  • Customized comfort and protection
  • Anti-fog, distortion-free eye shield ensures clear vision
  • Latex-free adjustable nosepiece and soft, strong ear loops
  • Made from premium materials; mask will not lint, tear, or shred
  • Made in USA
SafeMask® Premier Elite™ ProShield earloop face masks provide ≥98% bacterial and particle filtration efficiency, the highest level of procedure mask protection. The fluid-resistant mask construction is also equipped with a unique shingle pleat design to prevent fluid pooling and cross-contamination.
Masks are constructed from a combination of spunbond Polypropylene (SBPP), melt blown Polypropylene (MBPP), Aluminum or Polypropylene (PP)-covered Iron, knitted Polyester and Spandex or knitted Polypropylene and Spandex.
Certifications: ASTM F 2100 Level 3 barrier protection.
Delivery information: Store in cool, clean, and dry conditions. Avoid excessive heat (over 40 °C or 104 °F).
Packaging: Masks are packaged 25 per box, 4 boxes per case.