Area Marking Tape Applicator
Supplier: Brady Worldwide
804314
77603-724EA 249.18 USD
77603-724
Area Marking Tape Applicator
Tapes
Easy-to-use machine lets you consistently and effectively lay down area marking tape in straight lines or curves faster than can by hand.
  • Steel
  • 3" core diameter
  • 2" maximum label width
Ergonomic design greatly reduces the need to bend down. Tension bar lets you adjust pressure on roller to increase tape adhesion to floor.