Rhodamine B 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
A951578-1G
A951578-25G
A951578-5G
A951578-100G
76729-306EA
8
USD
76729-306
76729-308
76729-310
77603-342
Rhodamine B 97%
Rhodamine B
Caution: for Research Use Only
Formula:
C₂₈H₃₁ClN₂O₃ |
MDL Number:
MFCD00011931 CAS Number: 81-88-9 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...