4-Chloro-7-nitrobenzofurazan 98%

Supplier: Ambeed
Warning

A880519-250MG A880519-25G A880519-5G A880519-1G A880519-100G
77036-782EA 17 USD
77036-782 77036-784 77036-786 77603-204 77882-801
4-Chloro-7-nitrobenzofurazan 98%
4-Chloro-7-nitrobenzofurazan

Caution: for Research Use Only

Formula: C₆H₂ClN₃O₃
MDL Number: MFCD00005808
CAS Number: 10199-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR