(S)-(+)-4-Chlorodinaphthol[2,1-d:1',2'-f][1,3,2]dioxaphosphepin ≥95%

Supplier: Ambeed
Danger

Synonyms: (S)-4-Chloro-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin

A816330-100MG A816330-250MG A816330-1G A816330-5G
77603-070EA 52.28 USD
77603-070 77603-072 77670-152 77873-280
(S)-(+)-4-Chlorodinaphthol[2,1-d:1',2'-f][1,3,2]dioxaphosphepin ≥95%
(S)-(+)-4-Chlorodinaphthol[2,1-d:1',2'-f][1,3,2]dioxaphosphepin

Caution: for Research Use Only

Formula: C₂₀H₁₂ClO₂P
MDL Number: MFCD02093048
CAS Number: 137156-22-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR