Tetramethylthiuram monosulfide ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Bis(dimethylthiocarbamyl) sulphide, Bis(dimethylthiocarbamyl) sulfide
A716104-100G
A716104-500G
A716104-25G
77602-834EA
12.3
USD
77602-834
77602-836
77669-922
Tetramethylthiuram monosulfide ≥98%
Tetramethylthiuram monosulphide
Caution: for Research Use Only
Formula:
C₆H₁₂N₂S₃ |
MDL Number:
MFCD00014870 CAS Number: 97-74-5 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...