Sakuranetin ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-5-Hydroxy-2-(4-hydroxyphenyl)-7-methoxychroman-4-one

A702347-10MG A702347-1MG A702347-5MG
77602-790EA 115.83 USD
77602-790 77602-792 77602-794
Sakuranetin ≥98%
Sakuranetin

Caution: for Research Use Only

Formula: C₁₆H₁₄O₅
MW: 286.29 g/mol
MDL Number: MFCD00075651
CAS Number: 2957-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR