Linagliptin 98%

Supplier: Ambeed
Warning

Synonyms: (R)-8-(3-Aminopiperidin-1-yl)-7-(but-2-yn-1-yl)-3-methyl-1-((4-methylquinazolin-2-yl)methyl)-1H-purine-2,6(3H,7H)-dione

A392251-10MG A392251-1G A392251-50MG A392251-5G A392251-5MG A392251-25G A392251-10G A392251-250MG
77033-558EA 7 USD
77033-558 77033-560 77033-562 77033-564 77033-566 77601-726 77668-816 77668-818
Linagliptin 98%
Linagliptin

Caution: for Research Use Only

Formula: C₂₅H₂₈N₈O₂
MDL Number: MFCD14635356
CAS Number: 668270-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR