N-Lauroylsarcosine sodium salt ≥98%

Supplier: Ambeed
Danger

A297457-25G
77601-328EA 6.15 USD
77601-328
N-Lauroylsarcosine sodium salt ≥98%
N-Lauroylsarcosine sodium salt

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MDL Number: MFCD00042728
CAS Number: 137-16-6
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR