Tristearin (glycerol tristearate) 97%

Supplier: Ambeed
Warning

Synonyms: Propane-1,2,3-triyl tristearate, Tristearin

A221844-1000G A221844-100G A221844-500G
77550-438EA 23 USD
77550-438 77568-138 77600-776
Tristearin (glycerol tristearate) 97%
Tristearin (Glycerol tristearate)

Caution: For research use only.

Formula: C₅₇H₁₁₀O₆
MDL Number: MFCD00036230
CAS Number: 555-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR