TAE684 ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-N4-(2-(isopropylsulphonyl)phenyl)-N2-(2-methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)pyrimidine-2,4-diamine, NVP-TAE 684, NVP-TAE684, 5-Chloro-N4-(2-(isopropylsulfonyl)phenyl)-N2-(2-methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)pyrimidine-2,4-diamine

A215396-100MG A215396-10MG A215396-250MG A215396-50MG
77600-740EA 453.05 USD
77600-740 77600-742 77600-744 77667-804
TAE684 ≥98%
TAE684

Caution: for Research Use Only

Formula: C₃₀H₄₀ClN₇O₃S
MDL Number: MFCD11977634
CAS Number: 761439-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR