Manganese(II) chloride 98%

Supplier: Ambeed
Danger

Synonyms: Manganous chloride, Manganese dichloride

A347892-100G
76773-256EA 5.31 USD
76773-256 76773-258 76773-260 77598-442
Manganese(II) chloride 98%
Manganese (II) chloride

Caution: for Research Use Only

Formula: MnCl₂
MW: 125.84 g/mol
Boiling Pt: 1190
Melting Pt: 650
MDL Number: MFCD00011114
CAS Number: 7773-01-5
UN: 3077
ADR: 9 Ⅲ
Merck Index: 12,05768

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR