Polyvinyl alcohol 2088

Supplier: Ambeed
Warning

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

A1495837-1000G A1495837-100G A1495837-500G
77597-710EA 13.33 USD
77597-710 77597-712 77622-254
Polyvinyl alcohol 2088
Polyvinyl alcohol

Caution: for Research Use Only

Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR