Polyethylene glycol monomethyl ether, M.W. ~1000

Supplier: Ambeed
Warning

Synonyms: Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH

A1460296-100G A1460296-25G A1460296-1000G A1460296-500G
77597-292EA 10.25 USD
77597-292 77597-294 77622-192 77867-818
Polyethylene glycol monomethyl ether, M.W. ~1000
Polyethylene glycol monomethyl ether

Caution: for Research Use Only

Formula: CH₃(OCH₂CH₂)nOH
MDL Number: MFCD00084416
CAS Number: 9004-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR