Alizarin complexone indicator

Supplier: Ambeed
Warning

A1456845-1G A1456845-25G A1456845-5G
77597-272EA 9.23 USD
77597-272 77622-178 77622-180
Alizarin complexone indicator
Alizarin complexone

Caution: for Research Use Only

Formula: C₁₉H₁₅NO₈
MDL Number: MFCD00001202
CAS Number: 3952-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR