Naphthalene black 10B (naphthol blue black, acid black 1) AR

Supplier: Ambeed
Warning

A1453416-100G
77597-244EA 19.48 USD
77597-244
Naphthalene black 10B (naphthol blue black, acid black 1) AR
Naphthalene Black 10B (Naphthol Blue Black, Acid Black 1)

Caution: for Research Use Only

Formula: C₂₂H₁₄N₆Na₂O₉S₂
MDL Number: MFCD00004017
CAS Number: 1064-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR