Polyethylene glycol 2,000
Supplier: Ambeed
|
Warning
|
A139063-1000G
A139063-100G
A139063-25G
A139063-500G
77597-114EA
20.5
USD
77597-114
77597-116
77597-118
77597-120
Polyethylene glycol 2,000
Polyethylene glycol 2,000
Caution: for Research Use Only
Formula:
H(OCH₂CH₂)nOH |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...