Sodium orthovanadate ≥99%

Supplier: Ambeed
Warning

A949923-1G A949923-25G A949923-5G
77595-532EA 6.15 USD
77595-532 77595-534 77595-536
Sodium orthovanadate ≥99%
Sodium orthovanadate

Caution: for Research Use Only

Formula: Na₃O₄V
MW: 183.91 g/mol
MDL Number: MFCD00003511
CAS Number: 13721-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR