N,N,N',N',N''-Pentamethyldiethylenetriamine 98%

Supplier: Ambeed
Danger

A877442-10G A877442-25G A877442-100G
77200-116EA 5 USD
77200-116 77308-482 77595-420
N,N,N',N',N''-Pentamethyldiethylenetriamine 98%
N,N,N',N',N''-Pentamethyldiethylenetriamine

Caution: for Research Use Only

Formula: C₉H₂₃N₃
MDL Number: MFCD00014876
CAS Number: 3030-47-5
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR