(±)-3-Amino-1,2-propanediol 95%

Supplier: Ambeed
Danger

A853353-25G A853353-100G
77171-654EA 5 USD
77171-654 77595-370
(±)-3-Amino-1,2-propanediol 95%
(±)-3-Amino-1,2-propanediol

Caution: for Research Use Only

Formula: C₃H₉NO₂
MDL Number: MFCD00008140
CAS Number: 616-30-8
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR