Zinc(II) dibutyldithiocarbamate ≥99%
Supplier: Ambeed
|
Danger
|
Synonyms:
Dibutyldithiocarbamic Acid Zinc(II) Salt
A550740-25G
A550740-100G
A550740-1000G
77594-682EA
5.13
USD
77594-682
77602-332
77872-923
Zinc(II) dibutyldithiocarbamate ≥99%
Zinc(II) dibutyldithiocarbamate
Caution: for Research Use Only
Formula:
C₉H₁₉NS₂ |
MDL Number:
MFCD00067274 CAS Number: 136-23-2 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...