Ammonium molybdate(VI) 99.9%

Supplier: Ambeed
Danger

A540893-25G
77594-666EA 65.6 USD
77594-666
Ammonium molybdate(VI) 99.9%
Ammonium molybdate(VI)

Caution: for Research Use Only

Formula: H₈MoN₂O₄
MDL Number: MFCD00011596
CAS Number: 13106-76-8
UN: 3288
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR