1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether ≥98%

Supplier: Ambeed
Danger

Synonyms: 2,2,3,3,4,4,5,5-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether, 1,1,2,2,3,3,4,4-Octafluoro-5-(1,1,2,2-tetrafluoroethoxy)pentane

A491001-25G
77594-532EA 64.58 USD
77594-532
1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether ≥98%
1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether

Caution: for Research Use Only

Formula: C₇H₄F₁₂O
MW: 332.09 g/mol
MDL Number: MFCD00155904
CAS Number: 16627-71-7
UN: 3271
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR