1,3,6-Trichloroisoquinoline 95%
Supplier: Ambeed
|
Danger
|
A389931-100MG
A389931-1G
A389931-250MG
A389931-5G
77115-630EA
12.36
USD
77115-630
77115-632
77115-634
77594-188
1,3,6-Trichloroisoquinoline 95%
1,3,6-Trichloroisoquinoline
Caution: for Research Use Only
Formula:
C₉H₄Cl₃N |
MDL Number:
MFCD09907620 CAS Number: 1053658-49-3 UN: 2923 ADR: 8,6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...