1-Bromo-4-(trifluoromethylthio)benzene 98%

Supplier: Ambeed
Danger

Synonyms: 4-Bromophenyl trifluoromethyl sulfide, 4-(Trifluoromethylthio)bromobenzene, 4-Bromophenyl trifluoromethyl sulphide

A343087-1G A343087-5G
77290-560EA 11 USD
77290-560 77594-006
1-Bromo-4-(trifluoromethylthio)benzene 98%
1-Bromo-4-(trifluoromethylthio)benzene

Caution: for Research Use Only

Formula: C₇H₄BrF₃S
MDL Number: MFCD00040840
CAS Number: 333-47-1
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR