2-Chloro-3-fluoropyridine 98%

Supplier: Ambeed
Warning

A151143-25G A151143-5G A151143-100G
76984-650EA 24 USD
76984-650 76984-652 77592-840
2-Chloro-3-fluoropyridine 98%
2-Chloro-3-fluoropyridine

Caution: for Research Use Only

Formula: C₅H₃ClFN
MDL Number: MFCD03095302
CAS Number: 17282-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR