Monarch® Spin DNA Gel Extraction Kits
Supplier: New England Biolabs (NEB)
T1120L T1120S
77591-840EA 422 USD
77591-840 77591-842
Monarch® Spin DNA Gel Extraction Kits
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Monarch® spin DNA gel extraction kit, a newer version of our original gel extraction kit, offers an efficient method for DNA gel extraction and purification, enabling up to 5 μg of high-quality DNA.
  • Achieve high yields (up to 5 μg) and high purity in the extraction of DNA from agarose gels
  • Elute in very small volumes, in as low as 5 μl for elution, for highly concentrated DNA
  • Column design that minimizes buffer retention and contaminant carry over
  • Optimized buffer system without the need to adjust pH or add isopropanol
  • Designed for sustainability with less plastic used in columns and kit compared to leading suppliers
This new version features upgraded spin columns precision-engineered to allow low elution, in as little as 5 μl, for highly concentrated DNA in downstream applications such as sequencing, restriction digestion, library construction, labeling and other enzymatic manipulations.
Our optimized buffer system allows efficient binding and purification of the DNA, without a need to monitor pH. These kits contain columns and bottles designed to significantly reduce plastic usage compared to leading suppliers, while maintaining structural integrity for centrifugation, vacuum manifolds and other steps.