Infant Heel Warmer
Supplier: Azer Scientific Inc.
ES23530
77591-830CS 114.49 USD
77591-830
Infant Heel Warmer
Incubators Infant Incubators
Infant heel warmers are designed to safely and comfortably increase blood flow to the infant's heel area. By increasing blood, the potential for painful bruising, infection, or cartilage damage caused by frequent or repeated heel sticks is minimized.
  • Designed and tested for infant safety
  • Conforms to infant's foot for even heat distribution
  • Trigger disk allows for quick and easy activation
  • Product is latex-free
A trigger disk allows for quick and easy activation. Heel warmers quickly and gently reach the optimal desired temperature of 105 °F.