Tuttnauer 2540ET Benchtop Autoclave with Closed Door Drying
Supplier: Heidolph NA, LLC
023210756
77591-716EA 6333.65 USD
77591-716
Tuttnauer 2540ET Benchtop Autoclave with Closed Door Drying
Sterilizers Autoclaves (Steam Sterilizers)
Digital display sterilizers are ideal for sterilizing wrapped and unwrapped solid, hollow, or porous loads.
  • Plug and play - no plumbing required and no need to connect to building utilities
  • Closed Door Drying
  • Internal memory - storing up to 500 cycles
  • Cycle programs for wrapped and unwrapped instruments, wrapped pouches and small loads
  • Connectivity: USB port (×2), Wifi compatibility, ethernet port (×1)
  • 3 year-warranty
  • 5 to 6 week lead time
  • Wastewater tank: 3.75 L
This 21 L tabletop autoclave boasts the simplicity and fundamentals needed for everyday sterilization. This plug-and-play unit requires no dedicated connections for installation, simply a power source. All units require the reservoir to be filled with mineral-free water and come with 4 trays. An optional thermal printer is available. Ideal for sterilizing glassware, wrapped instruments, and unwrapped instruments.
Certifications: CE certified.
Ordering information: Stainless steel wire baskets and containers available for all autoclave models (sold separately). Optional printer available (sold separately).