Scepter™ Cut-Resistant Glove Liner with 3 Half Fingers
Supplier: Wells Lamont
M123
77591-714PK 81.59 USD
77591-714
Scepter™ Cut-Resistant Glove Liner with 3 Half Fingers
Gloves
This one-size-fits-all, cut-resistant glove is constructed of stainless steel and polyester, providing comfort and protection on the job.
  • Cut-resistant
  • Antimicrobial
  • 3 half fingers
  • Latex-free
This cut-resistant glove liner is designed with 3 half fingers. The thumb, index, and middle fingers are half fingers, ensuring greater dexterity and tactile sensation.
Certifications: Meets ANSI A4 for cut resistance.