Felt Pens, Alcohol-resistant
Supplier: SARSTEDT INC
95.950
77589-994CS 2181.97 USD
77589-994
Felt Pens, Alcohol-resistant
Pens Lab Markers
Hiqh quality labware from SARSTEDT.
  • Permanent labeling of many surfaces used in the laboratory
  • Alcohol-resistant on polystyrene and polycarbonate surfaces (e.g. cell culture dishes)
  • Limited suitability for polypropylene surfaces
  • For dry, wet and cold surfaces
  • Fine tip
Waterproof and fast-drying felt markers from SARSTEDT are perfect for non-smear labeling of plastic tubes and reaction tubes.