Tube Segment Opener
Supplier: SARSTEDT INC
95.1000 95.1000.476 95.1000.800
77589-928CS 638.75 USD
77589-928 77589-930 77589-932
Tube Segment Opener
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
SARSTEDT transfusion products are user-friendly, efficient and safe.
  • Description: Tube segment opener
  • Material: HDPE
  • Very easy sample collection from tube segments
  • Minimizes the risk of contamination
Safety and hygiene for the cross-matching laboratory.