Sealing Clips
Supplier: SARSTEDT INC
88.7708.095
77589-604CS 12051 USD
77589-604
Sealing Clips
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
SARSTEDT transfusion products are user-friendly, efficient and safe.
  • Stripper/sealer provides easy handling thanks to the ergonomically designed handle
  • Made of aluminium
  • Adjustable to different tube diameters
For manual stripping and sealing of blood bags.