Aspiration Pipette
Supplier: SARSTEDT INC
86.1252.011
77589-552CS 331.03 USD
77589-552
Aspiration Pipette
Pipettes Aspirating Pipettes
Dosing, transferring, and measuring liquids are routine procedures in almost every laboratory. SARSTEDT offers a wide range of products for every application, from high-quality pipette tips and serological pipettes to transfer pipettes, for reliable and reproducible results.
  • Working volume: 2 ml
  • Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
  • Polystyrene material
  • Without graduations
The Aspiration Pipette is designed for transferring liquid with no need for precision, and for the aspiration of liquids using a vacuum pump. This pipette is designed without print and supplied without cotton plug.