Transfer Pipette, 0.75 ml, with Graduations
Supplier: SARSTEDT INC
86.1170.300
77589-496CS 213.4 USD
77589-496
Transfer Pipette, 0.75 ml, with Graduations
Pipettes Pasteur/Transfer Pipettes
Dosing, transferring, and measuring liquids are routine procedures in almost every laboratory. SARSTEDT offers a wide range of products for every application, from high-quality pipette tips and serological pipettes to transfer pipettes, for reliable and reproducible results.
  • Suitable for transferring liquid diagnostic sample material
  • 0.1 ml graduations from 0.1 to 1.0 ml
  • Made of polyethylene without any environmentally harmful substances added
  • No risk of injury or infection as it is unbreakable
  • Suction bulb can be easily squeezed to ensure effortless pipetting
SARSTEDT transfer pipettes are essential for the safe and practical transfer of liquids in laboratories. The right pipette type for a variety of applications can be found in this standard product range.
Certifications: CE-certified in vitro diagnostic device.