Syringe Filters

Supplier: SARSTEDT INC
83.1826 83.1826.001 83.1826.102
77589-226CS 1576.04 USD
77589-226 77589-228 77589-230
Syringe Filters
Tubes Culture Tubes
To keep cells happy, the selection of a suitable cultivation surface is essential. SARSTEDT offers a wide range of cell culture labware and accessories for many applications.

  • Material: MBS
  • Connection: Luer lock
  • Membrane diameter: 28 mm
  • Filtration surface: 6.2 cm²
  • Purity: Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
  • Syringe filter Filtropur S for filtering cell culture media, cell culutre additives and buffers

PES membrane with 2 pore sizes: 0.45 µm for ultracleaning and clear filtration, 0.2 µm for sterile filtration; no evidence of protein absorption and a high flow rate.

Alternatively: Cellulose acetate (CA) membrane (pore size: 0.2 µm) with glass fiber (GF) pre-filter, for sterile filtration and increasing the total filtration volume.

SARSTEDT Filtropur offers filtration products for various volumes and applications. The range includes Filtropur V for vacuum filtration and syringe filters Filtropur S and Filtropur L for pressure filtration. This range is ideal for filtering aqueous solutions such as cell culture media.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR