Petri Dishes 100×100×20 mm, Square

Supplier: SARSTEDT INC
82.9923.422
77589-224CS 306.69 USD
77589-224
Petri Dishes 100×100×20 mm, Square
Culture Dishes Petri Dishes
One aspect of microbiology is researching and identifying pathogens, as well as looking for suitable treatments. SARSTEDT provides researchers with the consumables they need to do their work.

  • Ø×height: 100×100×20 mm
  • Ventilation cams: Yes
  • Often used for botanical analyses and applications due to the large diameter
  • Petri dishes with 6 ventilation cams in the lid for optimal gas exchange and reduced condensation
  • Fully automated production from crystal-clear polystyrene
  • Resistant to applications with approx. 60 °C hot and liquid agar
  • Sterilized by gamma irradiation

For the cultivation of microorganisms, SARSTEDT offers Petri dishes made of high-quality, crystal-clear polystyrene. Due to their high dimensional stability, they are ideal for use in automatic plate filling systems and are easily stacked.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR