Lid for ELISA and Microtest Plates
Supplier: SARSTEDT INC
82.1584
77589-210CS 117.83 USD
77589-210
Lid for ELISA and Microtest Plates
Lids Microplate Lids
Lids are suitable for use with ELISA and microtest plates in 96-well format.
  • Transparent lid made from polystyrene
  • Suitable for microtest and ELISA plates
  • With condensation rings to reduce evaporation
These lids are free from DNA/DNase/RNase, free from pyrogens and endotoxins, and non-cytotoxic.