Feces Tubes, with Special Feces Spoon
Supplier: SARSTEDT INC
80.623.022
77589-124CS 621.61 USD
77589-124
Feces Tubes, with Special Feces Spoon
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Faeces Sample Collection
In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, SARSTEDT offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
  • Material: PP
  • Label: With paper label
  • Design: With enclosed stirring rod
  • Particularly liquid and odor-tight
  • Secure screw cap for sending samples
  • The particular design of the spoon in the tube enables centrifugation and collection of a supernatant without disturbing it when opening the tube
In order to hygienically collect stool samples, all collection vessels are fitted with integral spoons. Fecal collection tubes combined with a broad and sturdy scoop in the screw cap make sampling easy.