Collection Cup, 125 ml
Supplier: SARSTEDT INC
75.565
77589-066CS 154.89 USD
77589-066
Collection Cup, 125 ml
Sample Containers General Purpose Containers
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Transparent polypropylene material
  • Break-proof and stackable
  • Graduated
  • Ideal height for test strips
SARSTEDT collection cups and containers are available in a range of sizes and volumes.