Urine Collection Cups
Supplier: SARSTEDT INC
75.1435.004
77589-044CS 126.69 USD
77589-044 77589-046
Urine Collection Cups
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, this range offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
  • Material: PP
  • Design: With safety label
  • Container with dual-click screw cap
  • Sterile, with safety label
  • Suitable for transport and storage of urine samples
  • Hygiene and convenience for patients and users
  • With graduations and labelling area
Small volume urine containers for consistent and reliable samples.