PCR Strips for qPCR - High Profile
Supplier: SARSTEDT INC
72.985 72.985.002 72.985.092 72.985.992
77589-006CS 291.48 USD
77589-006 77589-008 77589-010 77589-012
PCR Strips for qPCR - High Profile
Tubes PCR Tubes
SARSTEDT PCR consumables are compatible with standard thermocyclers. Uniform and extremely thin well walls facilitate consistently homogeneous and rapid heat transfer. During production, each individual well is tested to ensure it is 100% leak-proof.
  • High profile design
  • Supplied without closures
  • Polypropylene material
  • Optimal heat transfer through thin-walled reaction chamber
  • Optimized white color variant for maximum detection sensitivity in qPCR
  • Additional color variants for optimal organization in the laboratory
SARSTEDT PCR tubes are 'PCR Performance Tested' certified and guaranteed to be DNA-free, free from DNase/RNase, and free from PCR inhibitors. For the absolutely reliable absence of DNA and other biomolecules, Biosphere® plus certified products are subjected to a validated decontamination procedure. Biosphere® plus products are also free from DNA, DNase/RNase, PCR inhibitors, ATP, non-pyrogenic/endotoxin-free and are supplied sterile. Assorted color option includes a mix of Red, Green, Blue, and Purple.