PCR Plates with Full Skirt - Low Profile
Supplier: SARSTEDT INC
72.1980.700 72.1980 72.1980.010 72.1980.201 72.1980.600
77588-622CS 539.62 USD
77588-622 77588-614 77588-616 77588-618 77588-620
PCR Plates with Full Skirt - Low Profile
Microplates PCR Plates
SARSTEDT PCR consumables are compatible with standard thermocyclers. Uniform and extremely thin well walls facilitate consistently homogeneous and rapid heat transfer. During production, each individual well is tested to ensure it is 100% leak-proof.
  • Volume: 0.1 ml
  • 96-well, low profile design
  • Polypropylene material
  • Optimized white color variant for maximum detection sensitivity in qPCR
  • For low-volume reactions and fast PCR
  • Constant well and plate uniformity minimizes data variability
  • Low DNA and protein-binding variants for highest sample recovery
SARSTEDT PCR plates are 'PCR Performance Tested' certified and guaranteed to be DNA-free, free from DNase/RNase, and free from PCR inhibitors. For the absolutely reliable absence of DNA and other biomolecules, Biosphere® plus certified products are subjected to a validated decontamination procedure. Biosphere® plus products are also free from DNA, DNase/RNase, PCR inhibitors, ATP, non-pyrogenic/endotoxin-free and are supplied sterile.